Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B46391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46391G 1 g
B4639-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46391G Supplier TCI America Supplier No. B46391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133057-83-7
Molecular Formula C13H12BFO3
MDL Number MFCD00994627
InChI Key DQPDLDQJMUGVGI-UHFFFAOYSA-N
PubChem CID 2737788

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 1 g
Synonym 4-benzyloxy-3-fluorophenylboronic acid, 3-fluoro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-fluorobenzeneboronic acid, 4-benzyloxy-3-fluorophenyl boronic acid, 4-benzyloxy-3-fluorophenyl boranediol, boronic acid, 3-fluoro-4-phenylmethoxy phenyl, pubchem5322
SMILES B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)F)(O)O
IUPAC Name (3-fluoro-4-phenylmethoxyphenyl)boronic acid
Molecular Weight (g/mol) 246.044
Formula Weight 246.04
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-3-fluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.