Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. B450425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B450425G 25 g
B45045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B450425G Supplier TCI America Supplier No. B450425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54589-41-2
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.346
MDL Number MFCD00017508
InChI Key NMPNTBQOLRXPGK-UHFFFAOYSA-N
Synonym 4-benzyloxybenzophenone, 4-benzyloxy benzophenone, 4-benzyloxy phenyl phenyl methanone, phenyl-4-phenylmethoxyphenyl methanone, phenyl 4-phenylmethoxy phenyl ketone, acmc-20amoo, 4-benzyl-oxybenzophenone, 4-benzyloxyphenyl-phenyl-methanone
PubChem CID 458193
IUPAC Name phenyl-(4-phenylmethoxyphenyl)methanone
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 25 g
Formula Weight 288.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.