Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Benzyloxycarbonylamino)butyric acid, 98%

Catalog No. AAH6685706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6685706 5 g
AAH6685714 25 g
AAH6685722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6685706 Supplier Thermo Scientific Chemicals Supplier No. H6685706

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

CAS: 5105-78-2 | C12H15NO4 | 237.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5105-78-2
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.26
MDL Number MFCD00055835
InChI Key STQMDRQJSNKUAW-UHFFFAOYSA-N
Synonym z-gamma-abu-oh, 4-benzyloxy carbonyl amino butanoic acid, n-cbz-gaba, carbobenzoxy-4-aminobutyric acid, n-cbz-4-aminobutanoic acid, z-gaba-oh, n-cbz-4-aminobutyric acid, 4-carboxyamino butyric acid n-benzyl ester, z-l-gamma-aminobutyric acid
PubChem CID 21184
IUPAC Name 4-{[(benzyloxy)carbonyl]amino}butanoic acid
SMILES OC(=O)CCCNC(=O)OCC1=CC=CC=C1

Specifications

Beilstein 1884426
Quantity 5 g
Formula Weight 237.26
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Benzyloxycarbonylamino)butyric acid
RTECSNumber ES8137200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.