Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyindole 98.0+%, TCI America™
SDP

Catalog No. B28111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28111G 1 g
B28115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28111G Supplier TCI America Supplier No. B28111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20289-26-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
InChI Key LJFVSIDBFJPKLD-UHFFFAOYSA-N
Synonym 4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
PubChem CID 88465
IUPAC Name 4-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3

Specifications

Melting Point 62°C
Quantity 1 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.