Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylpyridine, 97+%

Catalog No. AAA1432622
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432622 100 g
AAA1432618 50 g
AAA1432636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1432622 Supplier Thermo Scientific Chemicals Supplier No. A1432622
Add to Cart

CAS: 2116-65-6 | C12H11N | 169.23 g/mol

4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2116-65-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006443
InChI Key DBOLXXRVIFGDTI-UHFFFAOYSA-N
Synonym pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
PubChem CID 16458
IUPAC Name 4-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=NC=C1

Specifications

Melting Point 9°C to 11°C
Density 1.061
Boiling Point 286°C to 287°C
Flash Point 115°C (239°F)
Refractive Index 1.5805
Quantity 100 g
Beilstein 115831
Solubility Information Soluble in ethanol,ethyl ether. Insoluble in water.
Formula Weight 169.23
Percent Purity 99%
Chemical Name or Material 4-Benzylpyridine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-319-0
RTECSNumber US2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.