Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B229425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2294-25G 25 g
B22945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2294-25G Supplier TCI America Supplier No. B229425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5122-94-1
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.03
MDL Number MFCD00093311
InChI Key XPEIJWZLPWNNOK-UHFFFAOYSA-N
PubChem CID 151253
IUPAC Name {[1,1'-biphenyl]-4-yl}boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 266°C
Color White-Yellow
Quantity 25 g
Synonym 4-biphenylboronic acid, 1,1 '-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid
Formula Weight 198.03
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.