Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenylyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. B50125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50125G 5 g
B50121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50125G Supplier TCI America Supplier No. B50125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17763-78-9
Molecular Formula C13H9F3O3S
Molecular Weight (g/mol) 302.267
MDL Number MFCD18379753
InChI Key RJFYGCJTUUXOOF-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 4-Biphenylyl Ester, 4-Biphenylyl Triflate
PubChem CID 11077493
IUPAC Name (4-phenylphenyl) trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 5 g
Formula Weight 302.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.