Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-ethylpyrazole 98.0+%, TCI America™
SDP

Catalog No. B51001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51001G 1 g
B51005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51001G Supplier TCI America Supplier No. B51001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71229-85-1
Molecular Formula C5H7BrN2
Molecular Weight (g/mol) 175.029
MDL Number MFCD08235252
InChI Key IPMSARLBJARXSC-UHFFFAOYSA-N
PubChem CID 12565201
IUPAC Name 4-bromo-1-ethylpyrazole
SMILES CCN1C=C(C=N1)Br

Specifications

Color Yellow
Boiling Point 108°C
Quantity 1 g
Formula Weight 175.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-ethylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.