Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC63901DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC63901DE 5 g
CC63901CB 250 mg
CC63901DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC63901DE Supplier Thermo Scientific Chemicals Supplier No. CC63901DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84547-86-4
Molecular Formula C5H5BrN2O2
Molecular Weight (g/mol) 205.011
InChI Key LEEPGDCCHVRYHK-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylicacid, pubchem16814, 4-bromo-1-methyl-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 536013
IUPAC Name 4-bromo-1-methylpyrazole-3-carboxylic acid
SMILES CN1C=C(C(=N1)C(=O)O)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.