Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-methylpyrazole 98.0+%, TCI America™
SDP

Catalog No. B406025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B406025G 25 g
B40605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B406025G Supplier TCI America Supplier No. B406025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15803-02-8
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.00
MDL Number MFCD02179565
InChI Key IXJSDKIJPVSPKF-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp
PubChem CID 167433
IUPAC Name 4-bromo-1-methyl-1H-pyrazole
SMILES CN1C=C(Br)C=N1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
Formula Weight 161.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-methylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.