Learn More
Sigma Organic Chemistry 4-Bromo-1H-pyrrole-2-ca | 1G | MFCD20502410
Supplier: Sigma Organic Chemistry CBR017801G

4-Bromo-1H-pyrrole-2-ca , 1G
About this Item:
MDL #: MFCD20502410
Molecular Weight: 170.99
UNSPSC Code: 12352200
Chemical Formula: C5H3BrN2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.