Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-2,2':6',2″-terpyridine 97.0+%, TCI America™
SDP

Catalog No. B32191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32191G 1 g
B32195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32191G Supplier TCI America Supplier No. B32191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149817-62-9
Molecular Formula C15H10BrN3
Molecular Weight (g/mol) 312.17
MDL Number MFCD06796965
InChI Key CJRRILXBSCRHKN-UHFFFAOYSA-N
PubChem CID 10614995
IUPAC Name 4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine
SMILES BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 1 g
Synonym 4′C-Bromo-alpha,alpha′C,alpha′C′C-tripyridyl
Formula Weight 312.17
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-2,2′:6′,2′′-terpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.