Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-dimethylaniline, 98%

Catalog No. AAA1071214
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1071214 25 g
AAA1071209 10 g
AAA1071218 50 g
AAA1071230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1071214 Supplier Thermo Scientific Chemicals Supplier No. A1071214
Add to Cart

CAS: 24596-19-8 | C8H10BrN | 200.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24596-19-8
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00007826
InChI Key QGLAYJCJLHNIGJ-UHFFFAOYSA-N
Synonym 4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl, 4-bromo-2,6-dimethyl-phenylamine, 2-amino-5-bromo-m-xylene, 2,6-dimethyl-4-bromoaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethyl-benzenamine
PubChem CID 90549
IUPAC Name 4-bromo-2,6-dimethylaniline
SMILES CC1=CC(Br)=CC(C)=C1N

Specifications

Melting Point 47°C to 52°C
Quantity 25 g
UN Number UN2811
Beilstein 1936300
Boiling Point 120°C (0.2 mmHg)
Formula Weight 200.08
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,6-dimethylaniline
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 246-337-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.