Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-dinitrotoluene, 97%, Thermo Scientific™

Catalog No. AC448860010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448860010 1 g Glass bottle
AC448860050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448860010 Supplier Thermo Scientific Chemicals Supplier No. 448860010
Add to Cart

Chemical Identifiers

CAS 95192-64-6
Molecular Formula C7H5BrN2O4
Molecular Weight (g/mol) 261.03
InChI Key UOGCLPDKGPPDHM-UHFFFAOYSA-N
Synonym 4-bromo-2,6-dinitrotoluene, 5-bromo-1,3-dinitro-2-methylbenzene, 5-bromo-2-methyl-1,3-dinitro-benzene, benzene, 5-bromo-2-methyl-1,3-dinitro, 2,6-dinitro-4-bromo-tolurene, 5-bromanyl-2-methyl-1,3-dinitro-benzene
PubChem CID 13443186
IUPAC Name 5-bromo-2-methyl-1,3-dinitrobenzene
SMILES CC1=C(C=C(C=C1[N+](=O)[O-])Br)[N+](=O)[O-]

Specifications

Melting Point 86°C to 88°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 261.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2, 6-dinitrotoluene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.