Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chloro-1-fluorobenzene 98.0+%, TCI America™
SDP

Catalog No. B186025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B186025G 25 g
B18605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B186025G Supplier TCI America Supplier No. B186025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60811-21-4
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00051794
InChI Key CJTIWGBQCVYTQE-UHFFFAOYSA-N
Synonym 1-bromo-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobromobenzene, 4-bromo-2-chlorofluorobenzene, 3-chloro-4-fluorobenzene bromide, 4-bromo-2-chloro-fluorobenzene, 4-bromo-2-chloro-1-fluoro-benzene, benzene, 4-bromo-2-chloro-1-fluoro, pubchem2169, acmc-209mma, ksc493o4n
PubChem CID 3543065
IUPAC Name 4-bromo-2-chloro-1-fluorobenzene
SMILES FC1=C(Cl)C=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
UN Number 3082
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-chloro-1-fluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.