Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluoro-N-methylbenzamide 98.0+%, TCI America™
SDP

Catalog No. B50741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50741G 1 g
B50745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50741G Supplier TCI America Supplier No. B50741G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 749927-69-3
Molecular Formula C8H7BrFNO
Molecular Weight (g/mol) 232.052
MDL Number MFCD09878362
InChI Key BAJCFNRLEJHPTQ-UHFFFAOYSA-N
PubChem CID 24694329
IUPAC Name 4-bromo-2-fluoro-N-methylbenzamide
SMILES CNC(=O)C1=C(C=C(C=C1)Br)F

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 232.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluoro-N-methylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.