Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. B195025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B195025G 25 g
B19505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B195025G Supplier TCI America Supplier No. B195025G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 112704-79-7
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00143263
InChI Key ZQQSRVPOAHYHEL-UHFFFAOYSA-M
Synonym 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754
PubChem CID 302681
IUPAC Name 4-bromo-2-fluorobenzoate
SMILES [O-]C(=O)C1=CC=C(Br)C=C1F

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 25 g
Formula Weight 219.01
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.