Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobiphenyl 99.0+%, TCI America™
SDP

Catalog No. B202825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B202825G 25 g
B20285G 5 g
B2028250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B202825G Supplier TCI America Supplier No. B202825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41604-19-7
Molecular Formula C12H8BrF
Molecular Weight (g/mol) 251.10
MDL Number MFCD00051716
InChI Key HTRNHWBOBYFTQF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
PubChem CID 521063
IUPAC Name 4-bromo-2-fluoro-1,1'-biphenyl
SMILES FC1=CC(Br)=CC=C1C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3152
Formula Weight 251.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-2-fluorobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.