Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. B183525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B183525G 25 g
B18355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B183525G Supplier TCI America Supplier No. B183525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2105-94-4
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 191.00
MDL Number MFCD00011722
InChI Key RYVOZMPTISNBDB-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene
PubChem CID 2724981
IUPAC Name 4-bromo-2-fluorophenol
SMILES OC1=CC=C(Br)C=C1F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.