Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-2'-hydroxyacetophenone, 95%

Catalog No. AAH61137MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61137MD 250 mg
AAH6113703 1 g
AAH6113706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61137MD Supplier Thermo Scientific Chemicals Supplier No. H61137MD

CAS: 30186-18-6 | C8H7BrO2 | 215.05 g/mol

4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30186-18-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD03428533
InChI Key LQCMMXGKEGWUIM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
PubChem CID 10727403
IUPAC Name 1-(4-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(Br)C=C1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 215.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4'-Bromo-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.