Learn More
4-Bromo-2-methoxybenzyl alcohol, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
It is used in azaquinone-methide-mediated depolymerization of aromatic carbamate oligomers.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 66°C to 70°C |
| Quantity | 1 g |
| Synonym | 4-bromo-2-methoxybenzyl alcohol, 4-bromo-2-methoxyphenyl methanol, 4-bromo-2-methoxy-phenyl-methanol, benzenemethanol, 4-bromo-2-methoxy, acmc-209e2a, 2-methoxy-4-bromobenzyl alcohol, benzenemethanol,4-bromo-2-methoxy |
| Solubility Information | Insoluble in water. |
| Formula Weight | 217.06 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Bromo-2-methoxybenzyl alcohol |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.