Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. AC451780500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451780500 50 g
AC451780100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451780500 Supplier Thermo Scientific Chemicals Supplier No. 451780500

Chemical Identifiers

CAS 139937-37-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.55
InChI Key KNBSFUSBZNMAKU-UHFFFAOYSA-N
Synonym 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072
PubChem CID 2781338
IUPAC Name 4-bromo-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 62.0°C to 67.0°C
Boiling Point 164.0°C (15.0 mmHg)
Assay Percent Range 96% min. (GC)
Linear Formula Br(CH3)C6H3SO2Cl
Quantity 50 g
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzene-1-sulfonyl chloride
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.