Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B38105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38105G 5 g
B381025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38105G Supplier TCI America Supplier No. B38105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99277-71-1
Molecular Formula C7H3BrNO4
Molecular Weight (g/mol) 245.01
MDL Number MFCD01013599
InChI Key ZIRHHEZLJGORGU-UHFFFAOYSA-M
Synonym 4-bromo-2-nitrobenzoic acid, 4-bromo-2-nitro-benzoic acid, 2-nitro-4-bromobenzoic acid, 5-bromo-2-carboxynitrobenzene, benzoic acid, 4-bromo-2-nitro, 4-bromo-2-nitrobenzoicacid, pubchem2055, 4-bromo-2-nitrobenzoic, acmc-209sc4, bromonitrobenzoicacid 42-
PubChem CID 3774467
IUPAC Name 4-bromo-2-nitrobenzoate
SMILES [O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 246.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.