Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-(trifluoromethoxy)aniline 98.0+%, TCI America™
SDP

Catalog No. B378625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B378625G 25 g
B37865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B378625G Supplier TCI America Supplier No. B378625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175278-09-8
Molecular Formula C7H5BrF3NO
Molecular Weight (g/mol) 256.02
MDL Number MFCD00179338
InChI Key QVILSWLYJYMGRN-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy aniline, 4-bromo-2-trifluoromethoxy-phenylamine, 2-amino-5-bromo trifluoromethoxy benzene, benzenamine, 4-bromo-2-trifluoromethoxy, 1-amino-4-bromo-2-trifluoromethoxy benzene, buttpark 50\01-74, 4-bromo-2-trifluoromethoxy phenylamine, pubchem2930, acmc-1c9j8, ksc184e5f
PubChem CID 737382
IUPAC Name 4-bromo-2-(trifluoromethoxy)aniline
SMILES NC1=CC=C(Br)C=C1OC(F)(F)F

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 256.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.