Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. TL00664DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00664DA 1 g Amber Glass Bottle
TL00664EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TL00664DA Supplier Thermo Scientific Chemicals Supplier No. TL00664DA
Add to Cart

Chemical Identifiers

CAS 175278-14-5
Molecular Formula C7H3BrClF3O3S
Molecular Weight (g/mol) 339.51
MDL Number MFCD00179336
InChI Key ZZCIRIWJHAZTIW-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy benzene-1-sulfonyl chloride, 4-bromo-2-trifluoromethoxy benzenesulfonyl chloride, 4-bromo-2-trifluoromethoxy benzene sulphonyl chloride, 4-bromo-2-trifluoromethoxy benzenesulfonylchloride, 4-bromo-2-trifluoromethoxy benzene, 4-bromo-2-trifluoromethoxy phenyl chlorosulfone, 4-bromo-2-trifluoromethoxy-benzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-trifluoromethoxy, benzenesulfonyl chloride, 4-bromo-2-trifluoromethoxy, 4-bromo-2-trifluoromethoxy benzenesulphonyl chloride
PubChem CID 2736421
IUPAC Name 4-bromo-2-(trifluoromethoxy)benzenesulfonyl chloride
SMILES FC(F)(F)OC1=C(C=CC(Br)=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Density 1.85g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 339.51
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.