Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-(trifluoromethyl)phenol 97.0+%, TCI America™
SDP

Catalog No. B449325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B449325G 25 g
B44935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B449325G Supplier TCI America Supplier No. B449325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50824-04-9
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD05668981
InChI Key PDPGERGWEOJVDC-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d
PubChem CID 142784
IUPAC Name 4-bromo-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)O

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 25 g
Formula Weight 241.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.