Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3,3,4,4-tetrafluoro-1-butene 98.0+%, TCI America™
SDP

Catalog No. B322225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B322225G 25 g
B32225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B322225G Supplier TCI America Supplier No. B322225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18599-22-9
Molecular Formula C4H3BrF4
Molecular Weight (g/mol) 206.97
MDL Number MFCD00042189
InChI Key GVCWGFZDSIWLMO-UHFFFAOYSA-N
Synonym 4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene
PubChem CID 87721
IUPAC Name 4-bromo-3,3,4,4-tetrafluorobut-1-ene
SMILES FC(F)(Br)C(F)(F)C=C

Specifications

Color Yellow
Boiling Point 60°C
Quantity 25 g
Formula Weight 206.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3,3,4,4-tetrafluoro-1-butene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.