Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3,5-dimethoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B295225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B295225G 25 g
B29525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B295225G Supplier TCI America Supplier No. B295225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31558-40-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00040799
InChI Key UGBJRYUNSXFPOX-UHFFFAOYSA-N
PubChem CID 3015579
IUPAC Name 4-bromo-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1Br)OC)C=O

Specifications

Melting Point 115°C
Quantity 25 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3,5-dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.