Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3,5-dimethylpyrazole 98.0+%, TCI America™
SDP

Catalog No. B330625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B330625G 25 g
B33065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B330625G Supplier TCI America Supplier No. B330625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3398-16-1
Molecular Formula C5H7BrN2
Molecular Weight (g/mol) 175.029
MDL Number MFCD00005242
InChI Key RISOHYOEPYWKOB-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylpyrazole, pyrazole, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl, 3.5-dimethyl-4-bromopyrazole, acmc-1cnce, 5-23-05-00154 beilstein handbook reference, ksc495g9b, 4-bromo-3,5-dimethyl pyrazole, 4-bromo-3,5-dimethyl-pyrazole
PubChem CID 76937
IUPAC Name 4-bromo-3,5-dimethyl-1H-pyrazole
SMILES CC1=C(C(=NN1)C)Br

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 25 g
Formula Weight 175.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3,5-dimethylpyrazole
RTECSNumber UQ6210000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.