Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-3'-chloroacetanilide 98.0+%, TCI America™
SDP

Catalog No. B314125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B314125G 25 g
B31415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B314125G Supplier TCI America Supplier No. B314125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22459-81-0
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040848
InChI Key BVADKOYGDKTWMU-UHFFFAOYSA-N
Synonym 4'-bromo-3'-chloroacetanilide, n-4-bromo-3-chlorophenyl acetamide, n-acetyl 4-bromo-3-chloroaniline, 4-bromo-3-chloroacetanilide, n1-4-bromo-3-chlorophenyl acetamide, acetamide,n-4-bromo-3-chlorophenyl, acmc-209fwl, maybridge1_008946, n-acetyl4-bromo-3-chloroaniline
PubChem CID 2746295
IUPAC Name N-(4-bromo-3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)Cl

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 248.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-3′-chloroacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.