Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-chloroaniline 98.0+%, TCI America™
SDP

Catalog No. B370925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B370925G 25 g
B37095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B370925G Supplier TCI America Supplier No. B370925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21402-26-6
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00041311
InChI Key QLYHPNUFNZJXOQ-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-3-chloro, 3-chloro-4-bromoaniline, unii-2gtm4sw4oh, 2gtm4sw4oh, 4-bromo-3-chloro-aniline, 4-bromo-3-chlorophenyl amine, pubchem3579, zlchem 1051, 4-bromo-3-chloro aniline, 4-bromo-3-chlorophenylamine
PubChem CID 88888
IUPAC Name 4-bromo-3-chloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Br

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.