Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-fluorotoluene 94.0+%, TCI America™
SDP

Catalog No. B193525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B193525G 25 g
B19355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B193525G Supplier TCI America Supplier No. B193525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-74-4
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00040940
InChI Key SLFNGVGRINFJLK-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562
PubChem CID 573280
IUPAC Name 1-bromo-2-fluoro-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)F

Specifications

Color Yellow
Boiling Point 191°C
Quantity 25 g
UN Number 1993
Formula Weight 189.03
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.