Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-methylbenzamide, 99%

Catalog No. AAL1617606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1617606 5 g
AAL1617603 1 g
AAL1617614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1617606 Supplier Thermo Scientific Chemicals Supplier No. L1617606
Add to Cart

CAS: 170229-98-8 | C8H8BrNO | 214.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170229-98-8
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.062
MDL Number MFCD00672914
InChI Key ZPFPOZMNEKPBIF-UHFFFAOYSA-N
Synonym benzamide,4-bromo-3-methyl, pubchem3804, 4-bromo-3-methyl-benzamide, acmc-1c5o5, 4-bromanyl-3-methyl-benzamide
PubChem CID 7020869
IUPAC Name 4-bromo-3-methylbenzamide
SMILES CC1=C(C=CC(=C1)C(=O)N)Br

Specifications

Melting Point 177°C to 182°C
Quantity 5 g
Beilstein 868074
Formula Weight 214.06
Percent Purity 99%
Chemical Name or Material 4-Bromo-3-methylbenzamide
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.