Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-4'-iodobiphenyl 98.0+%, TCI America™
SDP

Catalog No. B364825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B364825G 25 g
B36485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B364825G Supplier TCI America Supplier No. B364825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105946-82-5
Molecular Formula C12H8BrI
Molecular Weight (g/mol) 359.004
InChI Key GWOAJJWBCSUGHH-UHFFFAOYSA-N
Synonym 4-bromo-4'-iodobiphenyl, 4-bromo-4'-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-iodo, 1-bromo-4-4-iodophenyl benzene, 4'-bromo-4-iodobiphenyl, 4-iodo-4'-bromobiphenyl, 4-bromo-4'-iodo-biphenyl, 4-bromo-4/'-iodobiphenyl, ksc164g2f
PubChem CID 10904452
IUPAC Name 1-bromo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)Br

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 25 g
UN Number 3152
Formula Weight 359.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-iodobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.