Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. B46611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46611G 1 g
B4661200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46611G Supplier TCI America Supplier No. B46611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 942615-32-9
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD23099415
InChI Key SXOUNESKHJIXNK-UHFFFAOYSA-N
Synonym 4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene
PubChem CID 59182836
IUPAC Name 4-bromo-9,9-dimethylfluorene
SMILES CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 273.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-9,9-dimethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.