Learn More
Cayman Chemical 4-bromo A23187 5mg
SDP
Supplier:  Cayman Chemical 204385 MG
Encompass_Preferred
The first described limonoid a group of compounds known to impart bitter flavor in citrus fruits reported to demonstrate anti-proliferative and proapoptotic activity on several cancer cell lines as well as pesticidal anti-malarial anti-microbial and anti-HIV activity
Catalog No. 50-298-1771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.