Learn More
Sigma Organic Chemistry 4-Bromo-N- 2-methylphen 1G
Supplier: Sigma Organic Chemistry OTV0002691G

4-Bromo-N- 2-methylphen , 1G
About this item:
MDL #: MFCD00783630
Chemical Formula: C14H12BrNO
Molecular Weight: 290.16
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.