Alfa Aesar™ 4-Bromo-N-methylaniline, 97%

Manufacturer: Alfa Aesar™ H5527306

Quantity 5g

 View more versions of this product

Catalog No. AAH5527306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6911-87-1
    Formula Weight 186.05
    MDL Number MFCD03030391
    Chemical Name or Material 4-Bromo-N-methylaniline

    Specifications

    Boiling Point 259° to 260°C
    Formula Weight 186.05
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.6130
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Density 1.482g/mL
    Physical Form Liquid
    Quantity 5g
    MDL Number MFCD03030391
    CAS 6911-87-1
    Health Hazard 1 Warning
    Chemical Name or Material 4-Bromo-N-methylaniline
    Assay 97%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit