Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N,N-diethylaniline 97.0+%, TCI America™
SDP

Catalog No. B302725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B302725G 25 g
B30275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B302725G Supplier TCI America Supplier No. B302725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2052-06-4
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD00013530
InChI Key NGYMZFJVHHKJQR-UHFFFAOYSA-N
Synonym p-bromo-n,n-diethylaniline, benzenamine, 4-bromo-n,n-diethyl, n,n-diethyl-p-bromoaniline, 4-n,n-diethylamino-bromobenzene, benzenamine,4-bromo-n,n-diethyl, p-n,nddiethylaniline, 4-bromodiethylaniline, 4-bromophenyldiethylamine, 4-bromophenyl diethylamine, 4-bromo-phenyl diethylamine
PubChem CID 16328
IUPAC Name 4-bromo-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)Br

Specifications

Color White-Yellow
Boiling Point 215°C
Quantity 25 g
Formula Weight 228.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-N,N-diethylaniline
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.