Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N,N-dimethylaniline 98.0+%, TCI America™
SDP

Catalog No. B0585100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0585100G 100 g
B058525G 25 g
B0585500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0585100G Supplier TCI America Supplier No. B0585100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 586-77-6
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00000093
InChI Key XYZWMVYYUIMRIZ-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene
PubChem CID 11465
IUPAC Name 4-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(Br)C=C1

Specifications

Melting Point 54°C
Color White
Boiling Point 264°C
Quantity 100 g
Formula Weight 200.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-N,N-dimethylaniline
RTECSNumber BW9300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.