Learn More
Sigma Organic Chemistry 4-Bromo-thiophen-2-yl - | 500mg | MFCD09475422
Supplier: Sigma Organic Chemistry CDS003130500MG
4-Bromo-thiophen-2-yl - , 500mg
About this Item:
MDL #: MFCD09475422
Molecular Weight: 202.07
UNSPSC Code: 12352200
Chemical Formula: C6H4BrNS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.