Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzotrifluoride, 99%

Catalog No. AAA1594214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1594214 25 g
AAA1594206 5 g
AAA1594222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1594214 Supplier Thermo Scientific Chemicals Supplier No. A1594214
Add to Cart

CAS: 402-43-7 | C7H4BrF3 | 225.008 g/mol

4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-43-7
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000398
InChI Key XLQSXGGDTHANLN-UHFFFAOYSA-N
Synonym 4-bromobenzotrifluoride, 1-bromo-4-trifluoromethyl benzene, benzene, 1-bromo-4-trifluoromethyl, p-bromobenzotrifluoride, 4-bromotrifluorotoluene, 4-bromo-alpha,alpha,alpha-trifluorotoluene, p-bromotrifluoromethylbenzene, 4-trifluoromethyl bromobenzene, 4-bromo-a,a,a-trifluorotoluene, 4-trifluoromethylbromobenzene
PubChem CID 67872
IUPAC Name 1-bromo-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Br

Specifications

Density 1.63
Boiling Point 154°C to 155°C
Flash Point 60°C (140°F)
Refractive Index 1.473
Quantity 25 g
UN Number UN1993
Beilstein 2045666
Solubility Information Not miscible or difficult to mix.
Formula Weight 225.01
Percent Purity 99%
Chemical Name or Material 4-Bromobenzotrifluoride
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-943-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.