Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. B0558250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0558250G 250 g
B055825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0558250G Supplier TCI America Supplier No. B0558250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 586-75-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000683
InChI Key DENKGPBHLYFNGK-UHFFFAOYSA-N
Synonym p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride
PubChem CID 68515
IUPAC Name 4-bromobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 38°C
Boiling Point 247°C
Quantity 250 g
UN Number 3261
Formula Weight 219.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzoyl Chloride
EINECSNumber (3)-0062
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.