Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromodibenzofuran 98.0+%, TCI America™
SDP

Catalog No. B43451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43451G 1 g
B43455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43451G Supplier TCI America Supplier No. B43451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89827-45-2
Molecular Formula C12H7BrO
Molecular Weight (g/mol) 247.091
MDL Number MFCD00185685
InChI Key DYTYBRPMNQQFFL-UHFFFAOYSA-N
PubChem CID 458256
IUPAC Name 4-bromodibenzofuran
SMILES C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)Br

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 1 g
Formula Weight 247.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromodibenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.