Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. B394925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B394925G 25 g
B39495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B394925G Supplier TCI America Supplier No. B394925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54446-36-5
Molecular Formula C12H10BrN
Molecular Weight (g/mol) 248.123
MDL Number MFCD07779504
InChI Key CCIVUDMVXNBUCY-UHFFFAOYSA-N
Synonym N-(4-Bromophenyl)aniline, 4-Bromo-N-phenylaniline
PubChem CID 6425786
IUPAC Name 4-bromo-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)Br

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 318°C
Quantity 25 g
Formula Weight 248.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromodiphenylamine
EINECSNumber (3)-0061
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.