Learn More
Medchemexpress LLC 4 -Bromoflavone 5g | 20525-20-6 | 301.13 g/mol | C15H9BrO2 | 5 G

Supplier: Medchemexpress LLC HY1157725G
4'-Bromoflavone is a brominated flavone used in biochemical research as a cancer chemopreventive agent and a potent inducer of phase II detoxification enzymes. It is supplied as a solid or as a DMSO solution for in vitro studies and is commonly used to study enzyme induction and xenobiotic metabolism.
- Induces phase II detoxification enzymes.
- Available as solid and as solution in DMSO.
- Multiple pack sizes available, including 250 mg, 1 g, 5 g, 10 g, and 50 g.
- High purity (97.7%) suitable for biochemical assays.
- Chemical formula C15H9BrO2 and molecular weight 301.13 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.