Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. B184725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B184725G 25 g
B18475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B184725G Supplier TCI America Supplier No. B184725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2567-29-5
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD00017869
InChI Key HZQLUIZFUXNFHK-UHFFFAOYSA-N
Synonym 4-bromomethylbiphenyl, 4-bromomethyl biphenyl, 4-bromomethyl-1,1'-biphenyl, 4-phenylbenzyl bromide, 1-bromomethyl-4-phenylbenzene, bromoethylbiphenyl, 4-biphenylmethylbromide, 4-bromomethyl-biphenyl
PubChem CID 257716
IUPAC Name 1-(bromomethyl)-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CBr

Specifications

Melting Point 85°C
Color White-Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 247.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.