Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenylhydrazine hydrochloride, 99%

Catalog No. p-76867
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162310100 10 g Glass bottle
AC162310500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162310100 Supplier Thermo Scientific Chemicals Supplier No. 162310100
Only null left
Add to Cart

CAS: 622-88-8 | C6H7BrN2·HCl | 223.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-88-8
Molecular Formula C6H7BrN2·HCl
Molecular Weight (g/mol) 223.51
MDL Number MFCD00012941
InChI Key RGGOWBBBHWTTRE-UHFFFAOYSA-N
Synonym 4-bromophenylhydrazine hydrochloride, 4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazine hcl, 4-bromo phenyl hydrazine hydrochloride, p-bromophenylhydrazine monohydrochloride, hydrazine, 4-bromophenyl-, monohydrochloride, 1-4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazinehydrochloride, p-bromophenylhydrazine hydrochloride
PubChem CID 12157
IUPAC Name (4-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Br.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4NHNH2·HCl
Quantity 10 g
Beilstein 15, 434
Solubility Information Solubility in water: soluble
Formula Weight 223.51
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylhydrazine hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
RTECSNumber MV0800000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.