Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromotetrahydro-2H-pyran 98.0+%, TCI America™
SDP

Catalog No. B56001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56001G 1 g
B56005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56001G Supplier TCI America Supplier No. B56001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25637-16-5
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD07371501
InChI Key IVBVKTPDEWDNRW-UHFFFAOYSA-N
PubChem CID 13349654
IUPAC Name 4-bromooxane
SMILES C1COCCC1Br

Specifications

Boiling Point 61°C
Quantity 1 g
Formula Weight 165.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromotetrahydro-2H-pyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.