Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromotetraphenylsilane 96.0+%, TCI America™
SDP

Catalog No. B32871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32871G 1 g
B3287200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32871G Supplier TCI America Supplier No. B32871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18737-40-1
Molecular Formula C24H19BrSi
Molecular Weight (g/mol) 415.405
InChI Key UDZSLJULKCKKPX-UHFFFAOYSA-N
PubChem CID 297506
IUPAC Name (4-bromophenyl)-triphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)Br

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 1 g
Formula Weight 415.41
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromotetraphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.